Anti-p-AKT Rabbit Polyclonal Antibody [clone: T308]
Catalog # BWRLAP0486
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-AKT
- Clonality:Polyclonal
- Clone:T308
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human AKT1 around the phosphorylation site of Threonine 308.
p-AKT (T308) pAb detects endogenous levels of AKT1 only when phosphorylated at Thr308. This antibody also recognizes AKT2 and AKT3 when phosphorylated at the corresponding residues.
Type: Primary
Antigen: p-AKT
Clonality: Polyclonal
Clone: T308
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat