Anti-MEK1 Rabbit Polyclonal Antibody [clone: P293]
Catalog # BWRLAP0402
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MEK1
- Clonality:Polyclonal
- Clone:P293
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 270-320 of Human MEK1.
MEK1 (P293) pAb detects endogenous levels of MEK1 protein.
Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone: P293
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat