Order Entry
Export
ContactUsLinkComponent
Anti-HDAC 10 Rabbit Polyclonal Antibody [clone: E24]
Anti-HDAC 10 Rabbit Polyclonal Antibody [clone: E24]
Catalog # BWRLBS1161
Anti-HDAC 10 Rabbit Polyclonal Antibody [clone: E24]
Catalog # BWRLBS1161

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HDAC 10
  • Clonality:
    Polyclonal
  • Clone:
    E24
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to the N-terminal of Human HDAC 10.

HDAC10 (E24) pAb detects endogenous levels of HDAC10 protein.

Type: Primary
Antigen: HDAC 10
Clonality: Polyclonal
Clone: E24
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat