Anti-Gastrin Rabbit Polyclonal Antibody [clone: M90pAb]
Catalog # BWRLBS1139
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Gastrin
- Clonality:Polyclonal
- Clone:M90pAb
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human Gastrin.
Gastrin (M90) pAb detects endogenous levels of Gastrin protein.
Type: Primary
Antigen: Gastrin
Clonality: Polyclonal
Clone: M90pAb
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat