Anti-ERalpha Rabbit Polyclonal Antibody [clone: L100]
Catalog # BWRLBS1114
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ER alpha
- Clonality:Polyclonal
- Clone:L100
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human ERalpha.
Estrogen Receptor-? (L100) pAb detects endogenous levels of Estrogen Receptor-? protein.
Type: Primary
Antigen: ERalpha
Clonality: Polyclonal
Clone: L100
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat