Anti-G3BP-1 Rabbit Polyclonal Antibody [clone: D226]
BWRLBS1132
- Antibody type:Primary
- Antigen symbol:G3BP-1
- Clonality:Polyclonal
- Clone:D226
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 196-246 of Human G3BP-1.
G3BP-1 (D226) pAb detects endogenous levels of G3BP-1 protein.
Type: Primary
Antigen: G3BP-1
Clonality: Polyclonal
Clone: D226
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse