Anti-Cyclin E1 Rabbit Polyclonal Antibody [clone: I119]
Catalog # BWRLBS1085
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cyclin E1
- Clonality:Polyclonal
- Clone:I119
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 90-135 of Human Cyclin E1.
Cyclin E1 (I119) pAb detects endogenous levels of Cyclin E1 protein.
Type: Primary
Antigen: Cyclin E1
Clonality: Polyclonal
Clone: I119
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat