Silicon dioxide 30% in water, colloidal dispersion, LUDOX® SM, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Density:1.22 g/ml (25 °C)
:M-Clarity™ quality level = MQ100
Specification Test Results
- Colour:Colourless or White
- Form:Liquid
- ICP Major Analysis (Silica component):Confirmed
- Specific gravity at 60 °F:1,209 - 1,227
- pH at 25 °C:9,3 - 10,3
- Viscosity at 25 °C:4,8 - 6,8 cps
- Silica:29,0 - 31,0%
- Ratio of SiO₂/Na₂O:45 - 56
- Size (Surface area):320 - 400 (m²/g)
- Sulphate (as Na₂SO₄):≤0,06%
- Transmission:≥74,0%
- Concentration:30 wt% suspension in H₂O