Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Prepared Media for Microbiology
Accessories for temperature logger
Accessories for temperature logger
Accessories for temperature logger
Catalog # MULL901.0001
Supplier:
PHARMAMEDIA DR MULLER
CAS Number:
Expand image
Accessories for temperature logger
Catalog # MULL901.0001
Supplier:
PHARMAMEDIA DR MULLER
CAS Number:
Specifications
Pk:
1
Description:
T-Logger, measures temperature during transport
Specifications
About this item
Product Details
Product Details