1,4-Pentanediol ≥98.5%, Sigma-Aldrich®
Catalog # SIAL194182-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Assay:99%
- Appearance (Color):Colorless to Very Light Yellow
- Appearance (Form):Liquid
- Infrared Spectrum:Conforms to Structure
- Purity (GC):≥98,5%
- Refractive Index:n20/D 1,447 (Lit.)
Specification Test Results
About this item
:
:104.149 g/mol
Boiling Pt:72...73 °C (0.03 mmHg)) (lit.)
Density:0.986 g/ml (25 °C) (lit.)
:M-Clarity™ quality level = MQ100
Flash Pt:>113 °C
CAS Number:626-95-9
Formula:C₅H₁₂O₂
MDL Number:MFCD00004560