Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dextran
Dextran, M.W. ~ 60,000 clinical grade
Dextran, M.W. ~ 60,000 clinical grade
Dextran, M.W. ~ 60,000 clinical grade
Supplier:
MP Biomedicals
Certificates
Expand image
Dextran, M.W. ~ 60,000 clinical grade
Supplier:
MP Biomedicals
About this item
CAS Number:
9004-54-0
:
Formula:
(C₆H₁₀O₅)n
MDL Number:
MFCD00130935