Alcaligenes sp. Choline Oxidase, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Activity: 10 units/mg solid or more (containing approx. 20% of stabilisers). Unit definition: One unit causes the formation of one micromole of hydrogen peroxide per minute at pH 8,0 at 37 °C. Ref.: P.J.G. Mann and J.H. Quastel, Biochem. J., 31: 869 (1937)
Lyophilised powder
Specifications
- CAS No.:9028-67-5
- Species:Alcaligenes sp.
- Storage conditions:−20 °C
- Form:Yellow lyophilized powder
- Enzyme name:Choline Oxidase
- Enzyme activity:>10 units/mg solid