Order Entry
Export
ContactUsLinkComponent
Aluminium oxide neutral, MP EcoChrom™ H15152
Aluminium oxide neutral, MP EcoChrom™ H15152
Catalog # ICNA0204654
Supplier:  MP Biomedicals
CAS Number:  
Aluminium oxide neutral, MP EcoChrom™ H15152
Catalog # ICNA0204654
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    100 kg
  • Seq:
    4553

Specifications

About this item

 :
 :101.96 g/mol
Density:3.94 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424