Order Entry
Export
ContactUsLinkComponent
Aluminium oxide, Ecochrom™ Alumina N Activity: I
Aluminium oxide, Ecochrom™ Alumina N Activity: I
Catalog # ICNA0202090
Supplier:  MP Biomedicals
CAS Number:  
Aluminium oxide, Ecochrom™ Alumina N Activity: I
Catalog # ICNA0202090
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
 :
 :101.96 g/mol
Density:3.94 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424