Disinfection trays
Supplier: BODE CHEMIE
Certificates
About this item
User-friendly PVC trays with lid.
- Available in different sizes
- Practical sieve made of white PVC
- Molded handles
These trays are used in preparation of disinfection solutions in all areas like hospitals and laboratories.