Solvent debubbler
Solvent debubbler with bracket, HPLC system
Catalog # REST25014
Supplier: Restek
Jump to:
Product Details & Documents
Bubbles in an LC system can cause check valve malfunctions and pump cavitation, seriously affecting pump performance. The debubbler removes bubbles from the fluid stream before it enters the pump.
- The gas/liquid interface is easily visible through the translucent wall
- Loosening the airtight cap releases the trapped gas
The debubbler features a PP body and bracket; universal inlet and outlet connecting tips are made of 316 stainless steel and accept 0,063", 0,085", and 0,125" ID tubing.
Specifications
- Description:Solvent debubbler with bracket
- For:HPLC system