Flame retardant overalls, MICROGARD® FR
Supplier: Ansell
Certificates
About this item
MICROGARD® FR is a flame retardant material designed to be worn over woven thermal protective garments such as NOMEX® or PYROVATEX®, offering protection from particulates and pressurised liquid spray without compromising worker protection in the event of a flash fire.
- Flame retardant treated Sontara/wood pulp/polyester fabric with good barrier to particulates and low level liquid spray
- Air and water vapour permeable (breathable) to help reduce the risk of heat stress
- Optimised body fit improves wearer comfort and safety
- Elasticated 2-piece hood, wrists, waist and ankles
- External overlocked, stitched seams
MICROGARD® FR offers wearers protection from liquid chemicals to EN Type 6 and particulates to EN Type 5, and peace of mind to workers in potentially explosive/flammable environments. To decrease the risk of burn injury wear over thermal protective workwear.
Typical applications include: Oil and petrochemicals, petroleum distribution and processing, utilities.
Certifications: Type 5/6, EN ISO 14116 (index 1/0/0), EN 1073-2, EN 1149-5; Cat. III
Caution: MICROGARD® FR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above.
Specifications
- Catalog No:
- ANSEBL95S-00111-07
- ANSEBL95S-00111-06
- ANSEBL95S-00111-05
- ANSEBL95S-00111-04
- ANSEBL95S-00111-03
- Model:
- 111
- 111
- 111
- 111
- 111
- Garment application:
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Risk area:
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Fabric:
- Sontara/wood pulp/polyester
- Sontara/wood pulp/polyester
- Sontara/wood pulp/polyester
- Sontara/wood pulp/polyester
- Sontara/wood pulp/polyester