PEEK tubing
: Restek
Made of PEEK (Polyetheretherketone).
- Less oxygen permeable and more temperature resistant than PTFE or Tefzel® tubing
- Use with PEEK finger tight or flangeless fittings
This inert PEEK tubing replaces stainless steel, titanium, PTFE or Tefzel® tubing.