Anti-ZNF268 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30611
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 268 Recombinant
- Antigen symbol:ZNF268
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:10795
- Storage buffer:In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
- Sequence:AQVPNQTCPNTVWKIDDLMDWHQENKDKLGSTAKSFECTTFGKLCLLSTKYLSRQKPHKCGTHGKSLKYIDFTSDYARNNPNGFQVHGKSFFHSKHEQTVIGIKYCESIESGKTVNKKSQL
- Storage temperature:Store at 4° C. For long term storage store at -20° C.
- Immunogen:Recombinant ZNF268.
- Purification:Antigen affinity purification
- Size:100 µL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant human ZNF268.
Type: Primary
Antigen: ZNF268
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human