Order Entry
Export
ContactUsLinkComponent
Anti-KRT7 Monoclonal Antibody [clone: rOV-TL12/30]
Anti-KRT7 Monoclonal Antibody [clone: rOV-TL12/30]
Catalog # ABNORAB00302
Supplier:  Abnova
Anti-KRT7 Monoclonal Antibody [clone: rOV-TL12/30]
Catalog # ABNORAB00302
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 7
  • Antigen symbol:
    KRT7
  • Clonality:
    Monoclonal
  • Clone:
    rOV-TL12/30
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3855
  • Storage buffer:
    In 10 mM PBS.
  • Storage temperature:
    Store at -20° C.
  • Immunogen:
    OTN 11 ovarian carcinoma cell line.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal antibody raised against human KRT7.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: rOV-TL12/30
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human