Anti-CRABP2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30673
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CRABP2 Recombinant
- Antigen symbol:CRABP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:1382
- Storage buffer:In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
- Sequence:IKQEGDTFYIKTSTTVRTTEINFKVGEEFEEQTVDGRPCKSLVKWESENKMVCEQKLLKGEGPKTSWTRELTNDGELILTMTADDVVCTRVYVRE
- Storage temperature:Store at 4° C. For long term storage store at -20° C.
- Immunogen:Recombinant CRABP2.
- Purification:Antigen affinity purification
- Size:100 µL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant human CRABP2.
Type: Primary
Antigen: CRABP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human