Anti-CDH2 Mouse Monoclonal Antibody [clone: 13A9]
ABNOMAB14461
: Abnova
- Antibody type:Primary
- Antigen name:Cadherin 2 Recombinant
- Antigen symbol:CDH2
- Clonality:Monoclonal
- Clone:13A9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:1000
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Storage temperature:Store at 4° C.
- Immunogen:Recombinant protein corresponding to cytoplasmic domain of human CDH2.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against partial recombinant human CDH2.
Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 13A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human