Specifications
- Pk:1
- Description:DNA Extraction Solution, QuickExtract™
Specifications
About this item
The QuickExtract™ DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from almost any sample type using a simple, one-tube protocol that takes only 3 to 8 minutes, depending on the sample. QuickExtract Solution has been used to extract DNA from samples such as hair follicles, quill-end cells of feathers, tissue-culture cells, buccal cells, zebrafish organs and scales, and mouse tail snips. The extracted DNA is suitable for PCR analysis, such as genomic, transgenic, or viral DNA screening in animals, or for genetic or environmental research and screening in humans and other organisms.
- Non-toxic reagents, inexpensive processing
- Short procedure (8 minutes for the average sample)
- No centrifugation or spin columns to reduce yields
- Compatible with high-throughput and robotic workflows
The QuickExtract™ method allows for the processing of one to hundreds of samples simultaneously, without centrifugation, spin columns or the use of any toxic organic solvent. The method is also compatible with robotic automation.