Orders LinkContactUsLinkComponent
Thermolite® septa
 :   Restek
Thermolite® septa
 :  Restek
Thermolite® septa
Thermolite® septa
These septa are preconditioned and ready to use.
  • Usable to 340 °C inlet temperature
  • Precision molding assures consistent, accurate fit
  • Excellent puncturability
Note: For 17 mm inlets, the maximum temperature for Thermolite septa is 290 °C.
 : Packaged in ultra-clean blister packs. 12,7 mm and 17 mm septa are packaged in precleaned glass jars.
Product Family Options
  • Description
    Ø
  • Thermolite® septa
    5 mm (3/16”) mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    5 mm (3/16”) mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    6 mm (1/4”) mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    6 mm (1/4”) mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    7 mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    7 mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    8 mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    8 mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    9 mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    9 mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    9,5 mm (3/8”) mm
  • Thermolite® septa
    9,5 mm (3/8”) mm
  • Thermolite® septa
    10 mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    10 mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    11,5 mm
    1 * 50 items
    1 * 50 items
  • Thermolite® septa
    11,5 mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    12,5 mm (1/2”) mm
  • Thermolite® septa
    12,5 mm (1/2”) mm
    1 * 100 items
    1 * 100 items
  • Thermolite® septa
    17 mm
    1 * 50 items
    1 * 50 items
Accessories
  •  
    Description
  • REST21299
    Septa holder kit with septum support and holder
    1 * 1 items - 137,00 €