Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rosiglitazone
Rosiglitazone ≥98%
Rosiglitazone ≥98%
:
Cayman Chemical
Rosiglitazone ≥98%
:
Cayman Chemical
:
5-(4-(2-(Methyl(pyridin-2-yl)amino)ethoxy)benzyl)thiazolidine-2,4-dione
:
122320-73-4
:
:
C₁₈H₁₉N₃O₃S
:
MFCD00871760
:
357.43 g/mol
Environmentally Preferable: