Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Supplier:   Merck
Glass beads
Supplier:  Merck
Certificates
2 Options Available Below
Product Details & Documents
Made of PYREX® glass, solid.
For boiling and distillation operations. Use as distillation packing, mixing beads or boiling stones.
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • SIAL18406-500G
    Glass beads
    Glass beads
    5 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,10 €
    57,10 €
    1 * 500 g
       57,10 €
  • Glass beads
    5 mm
    Log in for availability
    Log in for availability
    1 * 3.000 g
    1 * 3.000 g - 308,00 €
    308,00 €
    1 * 3.000 g
       308,00 €