Anti-HDGF Rabbit Polyclonal Antibody
Catalog # BSBTA01057
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:HDGF
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Hepatoma-derived growth factor(HDGF) detection. Tested with WB, IHC-P in Human, Mouse, Rat.
Type: Primary
Antigen: HDGF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat