Fitting kits, Swagelok®
Swagelok® fitting kit, Brass
Catalog # REST23141
Supplier: Restek
Jump to:
Product Details & Documents
This Swagelok® fitting kits includes the most common assortment of ¹/₈" and ¹/₄" brass fittings.
- Parts list makes reordering easy
- Parts come in sturdy toolbox for easy and convenient storage
Delivery information: The kit includes ¹/₈" brass nut (20), ¹/₄" brass nut (20), ¹/₈" brass front ferrule (20), ¹/₄" brass front ferrule (20), ¹/₈" brass back ferrule (20), ¹/₄" brass back ferrule (20), ¹/₈" brass cap (6), ¹/₄" brass cap (6), ¹/₈" brass plug (6), ¹/₄" brass plug (6), ¹/₈" brass union (2), ¹/₄" brass union (2), ¹/₄ to ¹/₈" brass reducing union (2), ¹/₈" brass tee (2), ¹/₄" brass tee (2), ¹/₈ to ¹/₄" brass tube end reducer (2), ¹/₄ to 1/8" brass tube end reducer (2), ¹/₈" inspection gauge (1) and ¹/₄" inspection gauge (1).
Specifications
- Description:Swagelok® fitting kit
- Material:Brass