Hot swap capillary column nuts
Catalog # REST22348
Supplier: Restek
Specifications
- Description:Hot swap capillary column nut, for use with compact Agilent-style ferrules
- Pk:1
Specifications
About this item
This hot swap capillary column nut allows to change capillary column while the injector temp is still hot without downtime waiting for injector parts to cool down.