Orders LinkContactUsLinkComponent
Capillary nuts
Supplier:   Restek
Capillary nuts
Supplier:  Restek
Certificates
Capillary nuts
2 Options Available Below
Product Details & Documents
These capillary nuts are ideal for Shimadzu 17A, 2010, and 2014 GCs.
The enhanced capillary nut design eliminates the slot, increasing lifetime and durability.
Product Family Options
Product Information
  • Availability
    Price
  • REST22688
    Capillary nut
    Capillary nut
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items - 83,00 €
    83,00 €
    1 * 2 items
       83,00 €
  • REST20375
    Enhanced capillary nut
    Enhanced capillary nut
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items - 82,00 €
    82,00 €
    1 * 2 items
       82,00 €