Injection port repair tool
Catalog # REST21393
Supplier: Restek
Specifications
- Description:Injection port repair tool
- Pk:1
Specifications
About this item
Injection port repair tool for agilent split/splitless injection ports.
- Remove contaminants from inside the injector to achieve a better seal
- Cleans critical inlet seal areas
- Saves time by preventing unwanted leaks
The sealing ring at the base of a split/splitless injector allows a seal to form between the injection port and the inlet disk. This sealing ring wears over time and may become scratched or pitted, which compromises the sealing ability of the injector to the inlet disk.
Delivery: Includes a handle and brush adaptor, 5 medium sanding disks, 5 fine sanding disks, one 6,5 mm bore brush, and one 7 mm bore brush.