Thioglycerol standard, 10 mM
Catalog # CAYM700643-100
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pk:100 µl
- Description:Thioglycerol standard
Specifications
About this item
The standard is 'ready to use' to prepare the standard curve in the lipase activity assay kit.
The vial contains 100 µl of 10 mM 1-thioglycerol.