Anti-CD5 Rat Monoclonal Antibody (FITC) [clone: 53-7.3]
Supplier: Tonbo Biosciences
Certificates
About this item
The 53-7.3 monoclonal antibody is specific for mouse CD5, a 67 kDa protein that is expressed on most thymocytes, mature T cells and the B-1 subset of B cells. CD5 is a member of the scavenger receptor cysteine-rich protein superfamily and is a ligand for the c-type lectin CD72. CD5 is also known as Leu-1 and functions to modulate antigen receptor signaling on T and B cells.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: FITC
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Specifications
- Catalog No:
- 35-0051-U025
- 35-0051-U100
- Antigen symbol:
- Ly-1, Ly-12, Ly-A, Lyt-1
- Ly-1, Ly-12, Ly-A, Lyt-1
- Antigen name:
- CD5
- CD5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 53-7.3
- 53-7.3
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 12507
- 12507
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- 2-8°C protected from light
- 2-8°C protected from light