Capillary installation gauges
Supplier: Restek
Jump to:
About this item
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
- Prevents crushed column ends
- Seats graphite ferrules onto column for consistent installations
4 Options Available Below
Product Details & Documents
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
- Prevents crushed column ends
- Seats graphite ferrules onto column for consistent installations
Product Family Options
Product Information
- AvailabilityPrice
- REST21034Capillary installation gauge for Agilent-style fittings (compact ferrules)Capillary installation gauge for Agilent-style fittings (compact ferrules)Log in for availability1 * 1 items
- REST21399Capillary installation gauge for 1/16" fittings (1/16" ferrules)
- REST22335Capillary installation gauge for "Bruker/Varian GCs for use with 1/16" ferrules"Capillary installation gauge for "Bruker/Varian GCs for use with 1/16" ferrules"Log in for availability1 * 1 items121,00 €
- REST22330Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)Log in for availability1 * 1 items119,00 €