Anti-OGDH/GLUD Mouse Monoclonal Antibody [clone: 9F4BD5]
Catalog # ABNOMAB13985
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:OGDH/GLUD Ogdh
- Antigen symbol:OGDH/GLUD
- Clonality:Monoclonal
- Clone:9F4BD5
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Storage buffer:In HBS (0.02% sodium azide).
- Immunogen:A mixture of native purified OGDH from porcine heart and native purified GLUD from bovine liver.
- Purification:Biochemical fractionation
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native OGDH subunit E2.
Type: Primary
Antigen: OGDH/GLUD
Clonality: Monoclonal
Clone: 9F4BD5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human