Anti-TOP1MT Mouse Monoclonal Antibody [clone: TOP1MT/488]
Catalog # ABNOMAB14239
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TOP1MT Recombinant
- Antigen symbol:TOP1MT
- Clonality:Monoclonal
- Clone:TOP1MT/488
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:116447
- Amino acid number:Q969P6
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Recombinant protein corresponding to full length human TOP1MT.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human TOP1MT.
Type: Primary
Antigen: TOP1MT
Clonality: Monoclonal
Clone: TOP1MT/488
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human