Anti-TNC Mouse Monoclonal Antibody [clone: T2H5]
Catalog # ABNOMAB14284
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:TNC
- Clonality:Monoclonal
- Clone:T2H5
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3371
- Amino acid number:P24821
- Storage buffer:In 1 mM PBS.
- Immunogen:Human breast carcinoma.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human TNC.
Type: Primary
Antigen: TNC
Clonality: Monoclonal
Clone: T2H5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human