Anti-THRA Mouse Monoclonal Antibody [clone: 2103]
ABNOMAB14178
: Abnova
- Antibody type:Primary
- Antigen symbol:THRA
- Clonality:Monoclonal
- Clone:2103
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:7067
- Amino acid number:P10827
- Storage buffer:In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
- Immunogen:A synthetic peptide corresponding to N-terminus of human THRA.
- Purification:Purified
- Size:100 µL
- Pk:100 µl
Mouse monoclonal antibody raised against synthetic peptide of human THRA.
Type: Primary
Antigen: THRA
Clonality: Monoclonal
Clone: 2103
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human