Inlet liner packing tool
Inlet liner packing tool, for liners ID ≥ 2 mm
Catalog # REST20339
Supplier: Restek
Jump to:
Product Details & Documents
The inlet liner packing tool can be used with all different liners.
- Accurate to a specific measured depth
- Position wool reproducibly every time
Specifications
- Description:Inlet liner packing tool, for liners ID ≥ 2 mm