Order Entry
Export
ContactUsLinkComponent
Human cathepsin B ELISA kit
Human cathepsin B ELISA kit
Catalog # BIRBORB50139-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human cathepsin B ELISA kit
Catalog # BIRBORB50139-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human Cathepsin B ELISA Kit
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <5 pg/ml

Specifications

About this item

Human cathepsin B ELISA kit is used for quantitative detection of human cathepsin B in sera, plasma, body fluids, tissue lysates or cell culture supernates.

Delivery: Kit includes lyophilised recombinant human cathepsin B standard, 2×10 ng/tube, one 96-well plate precoated with anti-human cathepsin B antibody, sample diluent buffer, 30 ml; biotinylated anti-human cathepsin B antibody, 130 µl; antibody diluent buffer, 130 µl; avidin-biotin-peroxidase complex (ABC), 12 ml; ABC diluent buffer, 12 ml; TMB colour developing agent, 10 ml; and TMB stop solution, 10 ml.