Encephalitis gE middle
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:Encephalitis gE middle
- Purity:Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining). Encephalitis protein was purified by proprietary chromatographic technique.
- Formulation:20 mM MES pH 6.5, 8M urea, 200 mM NaCL and 0.05% Tween-20.
- Tested applications:SDS-PAGE , HPLC , FuncS