Order Entry
Export
ContactUsLinkComponent
HCV Core-24
HCV Core-24
Supplier:  Biorbyt
Certificates
HCV Core-24
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HCV Core-24
  • Purity:
    HCV Core 24 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV-Core 24 protein was purified by proprietary chromatographic technique.
  • Formulation:
    20 mM Tris-Hcl pH 8, 1.5M urea and 10mM beta mercaptoethanol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information