Order Entry
Export
ContactUsLinkComponent
 
Dengue envelope-1 22 kDa
 :  Biorbyt
 
Dengue envelope-1 22 kDa
 :  Biorbyt

 

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Dengue Envelope-1 22kDa
  • Purity:
    Protein is >95% pure as determined by 10% PAGE (coomassie staining).
  • Sequence:
    TEAKQPATLR KYCIEAKLTN TTTDSRCPTQ GEPSLNEEQD KRFVCKHSMV DRGWGNGCGL FGKGGIVTCA MFTCKKNMKG KVVQPENLEY TIVITPHSGE EHAVGNDTGK HGKEIKITPQ SSITEAELTG YGTVTMECSP RTGLDFNEMV LLQMENKAWL VHRQWFLDLP LPWLPGADTQ GSNWIQKETL VTFKNPHAKK QDVVVLGSQE GAMHTALTGA TEIQMSSGNL LFTGHLKCRL RMDKLQLKG.
  • Formulation:
    50mM sodium chloride-phosphorous buffer, pH-7.5.
  • Tested applications:
    SDS-PAGE