GAD-65
BIRBORB86025-0.5
: Biorbyt
:
- Pk:0,5 mg
- Conjugation:Unconjugated
- Storage conditions:Aliquot and store at −20 °C, avoid repeated freeze/thaw cycles.
- Protein synonyms:glutamate decarboxylase
- Protein/peptide name:GAD-65
- Tested applications:SDS-PAGE