Mouse recombinant CNTF (from E. coli)
Supplier: Biorbyt
About this item
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- BIRBORB107698-5
- BIRBORB107698-25
- Protein/peptide name:
- CNTF
- CNTF
- Protein synonyms:
- Murine Ciliary Neurotrophic Factor
- Murine Ciliary Neurotrophic Factor
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- Sequence:
- AFAEQSPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNISLD SVDGVPVASTDRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHTLTLQVSAFAY QLEELMALLE QKVPEKEADG MPVTIGDGGLFEKKLWGLKVLQELSQWTVR SIHDLRVISS HHMGISAHES HYGAKQM
- AFAEQSPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNISLD SVDGVPVASTDRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHTLTLQVSAFAY QLEELMALLE QKVPEKEADG MPVTIGDGGLFEKKLWGLKVLQELSQWTVR SIHDLRVISS HHMGISAHES HYGAKQM
- Purity:
- > 95% by SDS-PAGE and HPLC analyses
- > 95% by SDS-PAGE and HPLC analyses
- Formulation:
- Lyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4, 2% trehalose
- Lyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4, 2% trehalose
- Molecular weight:
- 22.4kDa
- 22.4kDa
- Storage conditions:
- Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Tested applications:
- SDS-PAGE , HPLC
- SDS-PAGE , HPLC
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.BIRBORB107698-5Protein/peptide nameCNTFProtein synonymsMurine Ciliary Neurotrophic FactorProtein/peptide typeRecombinantSpeciesMouseSourceE. coliSequenceAFAEQSPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNISLD SVDGVPVASTDRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHTLTLQVSAFAY QLEELMALLE QKVPEKEADG MPVTIGDGGLFEKKLWGLKVLQELSQWTVR SIHDLRVISS HHMGISAHES HYGAKQMPurity> 95% by SDS-PAGE and HPLC analysesFormulationLyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4, 2% trehaloseMolecular weight22.4kDaStorage conditionsStore at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.Tested applicationsSDS-PAGE , HPLC - Specifications:
More
Specifications
Cat. No.BIRBORB107698-25Protein/peptide nameCNTFProtein synonymsMurine Ciliary Neurotrophic FactorProtein/peptide typeRecombinantSpeciesMouseSourceE. coliSequenceAFAEQSPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNISLD SVDGVPVASTDRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHTLTLQVSAFAY QLEELMALLE QKVPEKEADG MPVTIGDGGLFEKKLWGLKVLQELSQWTVR SIHDLRVISS HHMGISAHES HYGAKQMPurity> 95% by SDS-PAGE and HPLC analysesFormulationLyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4, 2% trehaloseMolecular weight22.4kDaStorage conditionsStore at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.Tested applicationsSDS-PAGE , HPLC



