0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
A/Brisbane/10/06
A/Brisbane/10/06
BIRBORB79777-1
:
Biorbyt
:
A/Brisbane/10/06
BIRBORB79777-1
:
Biorbyt
:
Pk:
1 mg
Storage conditions:
Store at −20 °C.
Protein/peptide name:
A/Brisbane/10/06
Purity:
Greater than 95% by SDS-PAGE gel analysis.