0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
H1N1 strain A/New Caledonia/20/99 IVR 116
H1N1 strain A/New Caledonia/20/99 IVR 116
BIRBORB79771-1
:
Biorbyt
:
H1N1 strain A/New Caledonia/20/99 IVR 116
BIRBORB79771-1
:
Biorbyt
:
Pk:
1 mg
Storage conditions:
Store at −20 °C.
Protein/peptide name:
H1N1 strain A/New Caledonia/20/99 IVR 116
Purity:
Greater than 95% by SDS-PAGE gel analysis.