Order Entry
Export
ContactUsLinkComponent
GH-20K
GH-20K
Catalog # BIRBORB97770-100
Supplier:  Biorbyt
CAS Number:  
GH-20K
Catalog # BIRBORB97770-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Storage conditions:
    Store at−20 °C.
  • Biological activity:
    3 U/mg
  • UniProtKB:
    O14643
  • Protein/peptide name:
    GH-20K
  • Purity:
    95% pure by SDS-PAGE and RP-HPLC
  • Solubility:
    It is recommended to reconstitute and dilute the lyophilizedPlacental-GH in 0.4% NaHCO3 adjusted to pH 11.0,which can then be further diluted to other aqueoussolutions. For long term storage it is recommended to adda carrier protein (0.1% HSA or BSA).
  • Molecular weight:
    20 kDa
  • Formulation:
    Lyophilized. Placental Growth Hormone protein waslyophilized from a 1 mg/ml solution with 0.0045 mMNaHCO3 and pH 11.0.

Specifications

About this item