HCV-NS3-22 kDa
Catalog # BIRBORB97934-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Storage conditions:Store at−20 °C.
- Protein/peptide name:HCV-NS3-22 kDa
- Purity:Greater than 95% by SDS-PAGE
- Formulation:Liquid. Supplied in 20 mM Tris-HCl, pH 8.0, 8 M ureaand 10 mM BME.
- Tested applications:ELISA , WB