HCV-NS4 19 kDa
Catalog # BIRBORB97947-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Storage conditions:Store at−20 °C.
- Protein/peptide name:HCV-NS4 19kDa
- Purity:Greater than 95% by SDS-PAGE
- Formulation:Liquid. Supplied in 25 mM Tris-HCl pH 7.5, 1 mM EDTA,1.5 M urea and 50% glycerol.
- Tested applications:ELISA , WB